Anti-IRX3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535149
CAPIPA535149EA 1327.13 CAD
CAPIPA535149
Anti-IRX3 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and Xenopus based on sequence homology. IRX3 is a member of the Iroquois homeobox gene family (seeIRX1; MIM 606197) and plays a role in an early step of neural development. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:10-1:50

Type: Primary
Antigen: IRX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR