Anti-SIX3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535166
CAPIPA535166EA 1857.5 CAD
CAPIPA535166
Anti-SIX3 Rabbit Polyclonal Antibody
This antibody is predicted to react with chicken and mouse based on sequence homology. This gene encodes a member of the sine oculis homeobox transcription factor family. The encoded protein plays a role in eye development. Mutations in this gene have been associated with holoprosencephaly type 2.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SIX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR