Anti-ZNF81 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535177
CAPIPA535177EA 1857.5 CAD
CAPIPA535177
Anti-ZNF81 Rabbit Polyclonal Antibody
This gene encodes a protein that likely functions as a transcription factor. The protein contains an N-terminal KRAB domain and several C2H2-type zinc finger motifs. Mutations in this gene cause mental retardation X-linked type 45 (MRX45).

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50

Type: Primary
Antigen: ZNF81
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR