Anti-CREB3L4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535178
CAPIPA535178EA 1774.17 CAD
CAPIPA535178
Anti-CREB3L4 Rabbit Polyclonal Antibody
cAMP response element-binding (CREB) proteins are a familyof mammalian transcription activators. For further backgroundinformation on CREB proteins, see CREB1 (MIM 123810).[supplied byOMIM].

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CREB3L4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR