Anti-ZNF155 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535184
CAPIPA535184EA 1857.5 CAD
CAPIPA535184
Anti-ZNF155 Rabbit Polyclonal Antibody
ZNF155 may be involved in transcriptional regulation.

  • Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50

Type: Primary
Antigen: ZNF155
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR