Anti-NKX6-3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535192
CAPIPA535192EA 1857.5 CAD
CAPIPA535192
Anti-NKX6-3 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. The NKX family of homeodomain proteins controls numerous developmental processes. Members of the NKX6 subfamily, including NKX6-3, are involved in development of the central nervous system (CNS), gastrointestinal tract, and pancreas.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NKX6-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR