Anti-RARB Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535206
CAPIPA535206EA 1960.37 CAD
CAPIPA535206
Anti-RARB Rabbit Polyclonal Antibody
This antibody is predicted to react with chicken and mouse based on sequence homology. This gene encodes retinoic acid receptor beta, a member ofthe thyroid-steroid hormone receptor superfamily of nucleartranscriptional regulators. This receptor localizes to thecytoplasm and to subnuclear compartments. It binds retinoic acid,the biologically active form of vitamin A which mediates cellularsignalling in embryonic morphogenesis, cell growth anddifferentiation. It is thought that this protein limits growth ofmany cell types by regulating gene expression. The gene was firstidentified in a hepatocellular carcinoma where it flanks ahepatitis B virus integration site. The gene expresses at least twotranscript variants; one additional transcript has been described,but its full length nature has not been determined.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50

Type: Primary
Antigen: RARB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR