Anti-ZKSCAN4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535208
CAPIPA535208EA 1857.5 CAD
CAPIPA535208
Anti-ZKSCAN4 Rabbit Polyclonal Antibody
ZKSCAN4 may be involved in the transcriptional activation of MDM2 and EP300 genes.

  • Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ZKSCAN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR