Anti-ZNF483 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535212
CAPIPA535212EA 1857.5 CAD
CAPIPA535212
Anti-ZNF483 Rabbit Polyclonal Antibody
ZNF483 may be involved in transcriptional regulation.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ZNF483
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR