Anti-TBX15 Rabbit Polyclonal Antibody

PA535214 PA535214
CAPIPA535214EA 885.75 CAD
10157-940 CAPIPA535214
Anti-TBX15 Rabbit Polyclonal Antibody
Probable transcriptional regulator involved in developmental processes.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50

Type: Primary
Antigen: TBX15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR