Anti-HSF2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535220
CAPIPA535220EA 1857.5 CAD
CAPIPA535220
Anti-HSF2 Rabbit Polyclonal Antibody
HSF2, as well as the related gene HSF1, encodes a protein that binds specifically to the heat-shock element and has homology to HSFs of other species. Heat shock transcription factors activate heat-shock response genes under conditions of heat or other stresses. Although the names HSF1 and HSF2 were chosen for historical reasons, these peptides should be referred to as heat-shock transcription factors.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR