Anti-PCGF6 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535222
CAPIPA535222EA 1774.17 CAD
CAPIPA535222
Anti-PCGF6 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and rat based on sequence homology. The protein encoded by this gene contains a RING finger motif, which is most closely related to those of polycomb group (PcG) proteins RNF110/MEL-18 and BMI1. PcG proteins are known to form protein complexes and function as transcription repressors. This protein has been shown to interact with some PcG proteins and act as a transcription repressor. The activity of this protein is found to be regulated by cell cycle dependent phosphorylation. Alternatively spliced transcript variants encoding different isoforms have been identified.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PCGF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR