Anti-FOXN3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535226
CAPIPA535226EA 1506.12 CAD
CAPIPA535226
Anti-FOXN3 Rabbit Polyclonal Antibody
This gene is a member of the forkhead/winged helixtranscription factor family. Checkpoints are eukaryotic DNAdamage-inducible cell cycle arrests at G1 and G2. Checkpointsuppressor 1 suppresses multiple yeast checkpoint mutationsincluding mec1, rad9, rad53 and dun1 by activating aMEC1-independent checkpoint pathway. Alternative splicing isobserved at the locus, resulting in distinct isoforms.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50

Type: Primary
Antigen: FOXN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR