Anti-ZNF654 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535229
CAPIPA535229EA 1857.5 CAD
CAPIPA535229
Anti-ZNF654 Rabbit Polyclonal Antibody
ZNF654 may be involved in transcriptional regulation.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ZNF654
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR