Anti-RELA Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535232
CAPIPA535232EA 1774.17 CAD
CAPIPA535232
Anti-RELA Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL(MIM 164910), RELA, or RELB (MIM 604758) to form the NFKB complex.The p50 (NFKB1)/p65 (RELA) heterodimer is the most abundant form ofNFKB. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA,MIM 164008 or NFKBIB, MIM 604495), which inactivate NFKB bytrapping it in the cytoplasm. Phosphorylation of serine residues onthe I-kappa-B proteins by kinases (IKBKA, MIM 600664, or IKBKB, MIM603258) marks them for destruction via the ubiquitination pathway,thereby allowing activation of the NFKB complex. Activated NFKBcomplex translocates into the nucleus and binds DNA atkappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or Gpurine; and Y is a C or T pyrimidine).

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50

Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR