Anti-TSC2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535235
CAPIPA535235EA 1857.5 CAD
CAPIPA535235
Anti-TSC2 Rabbit Polyclonal Antibody
Mutations in this gene lead to tuberous sclerosis complex.Its gene product is believed to be a tumor suppressor and is ableto stimulate specific GTPases. The protein associates with hamartinin a cytosolic complex, possibly acting as a chaperone forhamartin. Alternative splicing results in multiple transcriptvariants encoding different isoforms.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TSC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR