Anti-CUL4B Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535239
CAPIPA535239EA 1774.17 CAD
CAPIPA535239
Anti-CUL4B Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. This gene is a member of the cullin family. The encodedprotein forms a complex that functions as an E3 ubiquitin ligaseand catalyzes the polyubiquitination of specific protein substratesin the cell. The protein interacts with a ring finger protein, andis required for the proteolysis of several regulators of DNAreplication including chromatin licensing and DNA replicationfactor 1 and cyclin E. Multiple transcript variants encodingdifferent isoforms have been found for this gene.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CUL4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR