Anti-FAM159A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535255
CAPIPA535255EA 1857.5 CAD
CAPIPA535255
Anti-FAM159A Rabbit Polyclonal Antibody
The exact function of this protein remains unknown.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: FAM159A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR