Anti-GPR58 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535263
CAPIPA535263EA 1857.5 CAD
CAPIPA535263
Anti-GPR58 Rabbit Polyclonal Antibody
Orphan receptor.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TAAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR