Anti-ECE2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535266
CAPIPA535266EA 1774.17 CAD
CAPIPA535266
Anti-ECE2 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine and mouse based on sequence homology. Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ECE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR