Anti-BLMH Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535267
CAPIPA535267EA 1857.5 CAD
CAPIPA535267
Anti-BLMH Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse, rat and rabbit based on sequence homology. The normal physiological role of BLM hydrolase is unknown, but it catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxamide bond of its B-aminoalaninamide moiety thus protecting normal and malignant cells from BLM toxicity.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: BLMH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR