Anti-CNO Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535268
CAPIPA535268EA 1857.5 CAD
CAPIPA535268
Anti-CNO Rabbit Polyclonal Antibody
The BLOC-1 complex is required for normal biogenesis of lysosome-related organelles, such as platelet dense granules and melanosomes. Plays a role in intracellular vesicle trafficking.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CNO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR