Anti-RNF166 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535283
CAPIPA535283EA 1327.13 CAD
CAPIPA535283
Anti-RNF166 Rabbit Polyclonal Antibody
This antibody is predicted to react with rat based on sequence homology.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RNF166
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR