Anti-GBP2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535285
CAPIPA535285EA 1774.17 CAD
CAPIPA535285
Anti-GBP2 Rabbit Polyclonal Antibody
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR