Anti-ST7L Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535289
CAPIPA535289EA 1857.5 CAD
CAPIPA535289
Anti-ST7L Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ST7L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR