Anti-NDUFA4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535296
CAPIPA535296EA 1857.5 CAD
CAPIPA535296
Anti-NDUFA4 Rabbit Polyclonal Antibody
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed to be not involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NDUFA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR