Anti-OR1L3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535299
CAPIPA535299EA 1960.37 CAD
CAPIPA535299
Anti-OR1L3 Rabbit Polyclonal Antibody
Odorant receptor (Potential).

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: OR1L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR