Anti-MDH1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535307
CAPIPA535307EA 2043.02 CAD
CAPIPA535307
Anti-MDH1 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, chicken, mouse, porcine and rat based on sequence homology. MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: MDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR