Anti-PAX3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535309
CAPIPA535309EA 1774.17 CAD
CAPIPA535309
Anti-PAX3 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. PAX3 probable is a transcription factor associated with development of alveolar rhabdomyosarcoma.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: PAX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR