Anti-TPT1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535332
CAPIPA535332EA 1857.5 CAD
CAPIPA535332
Anti-TPT1 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, porcine and rabbit based on sequence homology. TPT1 is involved in calcium binding and microtubule stabilization.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: TPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR