Anti-CCHCR1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535335
CAPIPA535335EA 1774.17 CAD
CAPIPA535335
Anti-CCHCR1 Rabbit Polyclonal Antibody
CCHCR1 may be a regulator of keratinocyte proliferation or differentiation.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: CCHCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR