Anti-RAP1A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535336
CAPIPA535336EA 1857.5 CAD
CAPIPA535336
Anti-RAP1A Rabbit Polyclonal Antibody
LRPAP1 interacts with LRP1/alpha-2-macroglobulin receptor and glycoprotein 330.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: RAP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR