Anti-AP2A2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535339
CAPIPA535339EA 1960.37 CAD
CAPIPA535339
Anti-AP2A2 Rabbit Polyclonal Antibody
Adaptins are components of the adaptor complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Alpha adaptin is a subunit of the plasma membrane adaptor. It binds polyphosphoinositides.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: AP2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR