Anti-USF2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535342
CAPIPA535342EA 1857.5 CAD
CAPIPA535342
Anti-USF2 Rabbit Polyclonal Antibody
This antibody is predicted to react with rat based on sequence homology. USF2 is a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. This protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:50-1:100

Type: Primary
Antigen: USF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR