Anti-KLC1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535349
CAPIPA535349EA 1857.5 CAD
CAPIPA535349
Anti-KLC1 Rabbit Polyclonal Antibody
This antibody is predicted to react with rat based on sequence homology. Kinesin is a microtubule-associated force-producing protein that may play a role in organelle transport. The light chain may function in coupling of cargo to the heavy chain or in the modulation of its ATPase activity.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: KLC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR