Anti-VPS26A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535357
CAPIPA535357EA 1857.5 CAD
CAPIPA535357
Anti-VPS26A Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, C. elegans, mouse, rat and Xenopus and zebrafish based on sequence homology. VPS26A is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: VPS26A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR