Anti-ANXA7 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535358
CAPIPA535358EA 1857.5 CAD
CAPIPA535358
Anti-ANXA7 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse and non-human primate based on sequence homology. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins.The Annexin VII gene contains 14 exons and spans approximately 34 kb of DNA. An alternatively spliced cassette exon results in two mRNA transcripts of 2.0 and 2.4 kb which are predicted to generate two protein isoforms differing in their N-terminal domain. ANXA7 is a protein with a molecular weight of approximately 51 kDa with a unique, highly hydrophobic N-terminal domain of 167 amino acids and a conserved C-terminal region of 299 amino acids. The latter domain is composed of alternating hydrophobic and hydrophilic segments. Structural analysis of the protein suggests that Annexin VII is a membrane binding protein with diverse properties, including voltage-sensitive calcium channel activity, ion selectivity and membrane fusion.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 0.5 mg/ml, Immunohistochemistry (Paraffin): 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: ANXA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR