Anti-C1Q B Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535369
CAPIPA535369EA 1492.04 CAD
CAPIPA535369
Anti-C1Q B Rabbit Polyclonal Antibody
C1QB is a major constituent of the human complement subcomponent C1q. C1q associates with C1r and C1s in order to yield the first component of the serum complement system. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: C1Q B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR