Anti-CD15 Mouse Monoclonal Antibody (DyLight® 488) [clone: MC-480]

Supplier: Pierce Biotechnology

MA1022D488
CAPIMA1022D488EA 1564.58 CAD
CAPIMA1022D488
Anti-CD15 Mouse Monoclonal Antibody (DyLight® 488) [clone: MC-480]
Antibodies
It has been successfully used in ICC/IF and flow cytometry applications on mouse and human samples. SSEA-1 (stage-specific embryonic antigen-1) is a cell surface antigen, expressed in murine embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.

This Antibody has been successfully used in ICC/IF and flow cytometry applications on mouse and human samples. SSEA-1 (stage-specific embryonic antigen-1) is a cell surface antigen, expressed in murine embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.

Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: MC-480
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR