To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Podocalyxin-Like
- ChIP:Yes
- Clonality:Monoclonal
- Clone:TRA-1-60
- Gel shift:Yes
- Host:Mouse
- Isotype:IgM
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:TRA-1-60
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:PCLP,PCLP-1,keratan sulfate antigen TRA1-60,Gp200,PC
- Storage buffer:Proprietary buffer with a proprietary stabilizer and no preservative
- Storage temperature:4 °C, do not freeze
- Concentration:1 mg/ml
- Immunogen:Human embryonal carcinoma cell line 2102Ep.
- Purification:Affinity chromatography - MBP
- Cat. no.:CAPIMA1023HRP
Specifications
About this item
It has been successfully used in Western blot applications on human samples. TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
This Antibody has been successfully used in Western blot applications on human samples. TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
Type: Primary
Antigen: TRA-1-60
Clonality: Monoclonal
Clone: TRA-1-60
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human