Anti-OCT4 Mouse Monoclonal Antibody (DyLight® 488) [clone: 9B7]

Supplier: Pierce Biotechnology

MA1104D488
CAPIMA1104D488EA 1657.38 CAD
CAPIMA1104D488
Anti-OCT4 Mouse Monoclonal Antibody (DyLight® 488) [clone: 9B7]
Antibodies
This Antibody detects Oct4/Pou5F1 in huIt has been successfully used in ICC/IF applications. Oct4 is a transcription factor which plays a key role in embryonic development, especially during early embryogenesis. Oct4 is one of the four YaIt has been linked to tumor formation.

This Antibody has been successfully used in ICC/IF applications. Oct4 is a transcription factor which plays a key role in embryonic development, especially during early embryogenesis. Oct4 is one of the four Yamaka factors, necessary for embryonic stem cell pluripotency. A translocation of this gene with the Ewing's sarcoma gene, t(6;22)(p21;q12), has been linked to tumor formation.

Type: Primary
Antigen: OCT4
Clonality: Monoclonal
Clone: 9B7
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR