Anti-GADD45A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512383
CAPIPA5-12383EA 2801.79 CAD
CAPIPA5-12383
Anti-GADD45A Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine based on sequence homology. GADD45A responds to environmental stresses by mediating activation of the p38/JNK pathway via MTK1/MEKK4 kinase. The GADD45A gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The DNA damage-induced transcription of this gene is mediated by both p53-dependent and -independent mechanisms.

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: GADD45A (growth arrest and DNA-damage-inducible, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR