Anti-FOXP3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512396
CAPIPA5-12396EA 2801.79 CAD
CAPIPA5-12396
Anti-FOXP3 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with non-human primate based on sequence homology. FOXP3 is a member of the forkhead/winged-helix family of transcriptional regulators. Defects in FOXP3 gene are the cause of immunodeficiency polyendocrinopathy, enteropathy, X-linked syndrome (IPEX), also known as X-linked autoimmunity-immunodeficiency syndrome.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR