Anti-RFC2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512398
CAPIPA5-12398EA 2561.45 CAD
CAPIPA5-12398
Anti-RFC2 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine based on sequence homology. The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins, proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also called activator 1, is a protein complex consisting of five distinct subunits of 145, 40, 38, 37, and 36.5 kD. RFC2 is the 40 kD subunit, which has been shown to be responsible for binding ATP. Deletion of RFC2 gene has been associated with Williams syndrome.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: RFC2 (replication factor C (activator 1) 2, 40kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR