Anti-PPAP2C Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512401
CAPIPA5-12401EA 2573.22 CAD
CAPIPA5-12401
Anti-PPAP2C Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine based on sequence homology. PPAP2C is a member of the phosphatidic acid phosphatase (PAP) family. PAPs convert phosphatidic acid to diacylglycerol, and function in de novo synthesis of glycerolipids as well as in receptor-activated signal transduction mediated by phospholipase D. This protein is similar to phosphatidic acid phosphatase type 2A (PPAP2A) and type 2B (PPAP2B). All three proteins contain 6 transmembrane regions, and a consensus N-glycosylation site. This protein has been shown to possess membrane associated PAP activity.

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: PPAP2C (phosphatidic acid phosphatase type 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR