Anti-U2AF1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512406
CAPIPA5-12406EA 2561.45 CAD
CAPIPA5-12406
Anti-U2AF1 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine and mouse based on sequence homology. U2AF1 belongs to the splicing factor SR family. U2 auxiliary factor, comprising a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the small subunit which plays a critical role in both constitutive and enhancer-dependent RNA splicing by directly mediating interactions between the large subunit and proteins bound to the enhancers.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: U2AF1 (U2 small nuclear RNA auxiliary factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR