Anti-RUNX1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA512409
CAPIPA5-12409EA 2801.79 CAD
CAPIPA5-12409
Anti-RUNX1 Rabbit Polyclonal Antibody
Antibodies
Core binding factor (CBF) is a heterodimeric transcription factor that binds to the core element of many enhancers and promoters. The RUNX1 protein represents the alpha subunit of CBF and is thought to be involved in the development of normal hematopoiesis.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: RUNX1 (runt-related transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR