Anti-DLG7 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512415
CAPIPA5-12415EA 2444.36 CAD
CAPIPA5-12415
Anti-DLG7 Rabbit Polyclonal Antibody
DLG7 is a potential cell cycle regulator that may play a role in carcinogenesis of cancer cells and mitotic phosphoprotein regulated by the ubiquitin-proteasome pathway. The protein is also a key regulator of adherens junction integrity and differentiation that may be involved in CDH1-mediated adhesion and signaling in epithelial cells.

Western Blot: 1:1000

Type: Primary
Antigen: DLG7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR