Anti-STIP1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512423
CAPIPA5-12423EA 2444.36 CAD
CAPIPA5-12423
Anti-STIP1 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, hamster, non-human primate and rat based on sequence homology. STIP1 mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: STIP1 (stress-induced phosphoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR