Anti-DCTPP1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512428
CAPIPA5-12428EA 2561.45 CAD
CAPIPA5-12428
Anti-DCTPP1 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and rat based on sequence homology.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: DCTPP1 (dCTP pyrophosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR